Learn More
Grainger Brady HMIG Label, Adhesive, White, 3-1/8 in x 1-1/8 in, 1,000/Pk.
Catalog No. NC9773460
Encompass_Preferred
These HMIG rectangular labels are designed for facility, hazard communication, and safety identification. The labels feature a semi-gloss finish and a permanent acrylic adhesive, allowing them to adhere effectively to most surfaces while enabling clean removal without residue. They include a dedicated write-on area to facilitate clear documentation of hazard information.
  • Write-on surface for custom entry of chemical name, manufacturer, and date
  • Permanent acrylic adhesive for reliable bonding
  • Rectangle shape with 3.125 in. x 1.13 in. dimensions
  • RoHS compliant to 2011/65/EU and 2015/863
  • Recommended for indoor use
Catalog No. NC9773460 Supplier Grainger Supplier No. 8NDC8
These HMIG rectangular labels are designed for facility, hazard communication, and safety identification. The labels feature a semi-gloss finish and a permanent acrylic adhesive, allowing them to adhere effectively to most surfaces while enabling clean removal without residue. They include a dedicated write-on area to facilitate clear documentation of hazard information.
  • Write-on surface for custom entry of chemical name, manufacturer, and date
  • Permanent acrylic adhesive for reliable bonding
  • Rectangle shape with 3.125 in. x 1.13 in. dimensions
  • RoHS compliant to 2011/65/EU and 2015/863
  • Recommended for indoor use