Learn More
Labelmaster Inc Standard Hazcom (HMIS) Label, 4'' x 4'', PVC-Free Film, 500/roll
SDP
Catalog No. NC1551963
Encompass
Our HMIS labels serve as a primary way to comply with OSHA's Hazard Communication Standard and have an easy-to-understand rating system to inform employees about the potential hazards of chemicals they might encounter in their workplace. Fill in these 4 x 4-in., high-gloss, weather-resistant, film right to know labels with the Chemical Identity; the Health, Flammability, Physical Hazard ratings; and the Personal Protection letter code for chemicals you use regularly. They are sold in rolls of 500.
Catalog No. NC1551963 Supplier Labelmaster Inc Supplier No. HMIS0007
Our HMIS labels serve as a primary way to comply with OSHA's Hazard Communication Standard and have an easy-to-understand rating system to inform employees about the potential hazards of chemicals they might encounter in their workplace. Fill in these 4 x 4-in., high-gloss, weather-resistant, film right to know labels with the Chemical Identity; the Health, Flammability, Physical Hazard ratings; and the Personal Protection letter code for chemicals you use regularly. They are sold in rolls of 500.