Learn More
Lonza Walkersville HMM, Hepatocyte Basal Medium 500 mL
Catalog No. NC9466813
Encompass
HMM Hepatocyte Maintenance Medium Sterility: Neg pH: 7.1-7.3 Osmolality: 325-341 mOsm/kg Used to maintain fresh plated human and animal hepatocytes in culture during testing. Media Only. Kit sold separate. NC9466815
Catalog No. NC9466813 Supplier Lonza Walkersville Supplier No. CC3197
HMM Hepatocyte Maintenance Medium Sterility: Neg pH: 7.1-7.3 Osmolality: 325-341 mOsm/kg Used to maintain fresh plated human and animal hepatocytes in culture during testing. Media Only. Kit sold separate. NC9466815