Learn More
Lonza Walkersville HMM SingleQuots Kit
Supplier:  Lonza Walkersville CC4192
Encompass
HMM Hepatocyte Maintenance Medium 1 kit SingleQuot> Kit (growth factors cytokines and supplements) Sterility: Neg pH: 7.1-7.3 Osmolality: 325-341 mOsm/kg Used to maintain fresh plated human and animal hepatocytes in culture during testing. Kit only. Media searchable under NC9466813
Catalog No. NC9466815