Learn More
Medchemexpress LLC HMMNI-d3 (hydroxy dimetridazole-d3) | 1015855-78-3 | 160.15 | 5 MG
SDP
Catalog No. 5000449392
Encompass_Preferred
HMMNI-d3 (Hydroxy Dimetridazole-d3) is a deuterium-labeled variant of HMMNI (Hydroxy dimetridazole), which itself is a hydroxy metabolite of Dimetridazol. Dimetridazole belongs to the nitroimidazole class of agents known for combating protozoan infections. HMMNI-d3 is intended for research use only. Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-49392 Supplier Medchemexpress LLC Supplier No. HYW008216S5MG
HMMNI-d3 (Hydroxy Dimetridazole-d3) is a deuterium-labeled variant of HMMNI (Hydroxy dimetridazole), which itself is a hydroxy metabolite of Dimetridazol. Dimetridazole belongs to the nitroimidazole class of agents known for combating protozoan infections. HMMNI-d3 is intended for research use only. Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS