Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC HMMNI (Hydroxy Dimetridazole) | 936-05-0 | 157.13 | 25 G
SDP

Catalog No. 5000448144
Encompass_Preferred

HMMNI, also known as Hydroxy dimetridazole, is a hydroxy metabolite derived from Dimetridazole, a nitroimidazole class drug. Dimetridazole is recognized for its effectiveness against protozoan infections. HMMNI itself demonstrates antiprotozoal activity, including the inhibition of Trichomonas foetus.

  • Molecular weight: 157.13
  • Formula: C5H7N3O3
  • CAS no.: 936-05-0
  • Appearance: solid
  • Color: off-white to yellow
  • Purity: 99.91%
  • Soluble in DMSO at 100 mg/mL (636.42 mM)
  • Exhibits antiprotozoal activity
  • Inhibits Trichomonas foetus with an ED50 of 1.6 μg/mL
  • Suitable for research use

Catalog No. 50-004-48144 Supplier Medchemexpress LLC Supplier No. 25GHYW008216
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

HMMNI, also known as Hydroxy dimetridazole, is a hydroxy metabolite derived from Dimetridazole, a nitroimidazole class drug. Dimetridazole is recognized for its effectiveness against protozoan infections. HMMNI itself demonstrates antiprotozoal activity, including the inhibition of Trichomonas foetus.

  • Molecular weight: 157.13
  • Formula: C5H7N3O3
  • CAS no.: 936-05-0
  • Appearance: solid
  • Color: off-white to yellow
  • Purity: 99.91%
  • Soluble in DMSO at 100 mg/mL (636.42 mM)
  • Exhibits antiprotozoal activity
  • Inhibits Trichomonas foetus with an ED50 of 1.6 μg/mL
  • Suitable for research use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.