Learn More
Medchemexpress LLC HY-13647 5mg Medchemexpress, HMN-176 CAS:173529-10-7 Purity:>98%
SDP
Catalog No. 502081444
Encompass_Preferred
Medchemexpress, HY-13647 5mg HMN-176 CAS:173529-10-7 HMN-176 is a stilbene derivative which inhibits mitosis, interfering with polo-like kinase-1 (plk1), without significant effect on tubulin polymerization.  Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1444 Supplier Medchemexpress LLC Supplier No. HY136475MG
Medchemexpress, HY-13647 5mg HMN-176 CAS:173529-10-7 HMN-176 is a stilbene derivative which inhibits mitosis, interfering with polo-like kinase-1 (plk1), without significant effect on tubulin polymerization.  Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.