Learn More
Innovative Research Inc Hmn ATP5PB Prtn Lysate 20ug
Catalog No. 502744355
Encompass_Preferred
Human Beta-2 Microglobulin (B2M) Purified 1mg - Human ATP5PB Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species Human Format Lyophilized Gene Symbol ATP5PB Gene Alias ATP5PB, PIG47, PIG47, ATP5F1 Protein Accession Number NP_001679 Nucelotide Accession Number NM_001688 Molecular Weight 28.9 kDa Storage Conditions -20C.
Catalog No. 50-274-4355 Supplier Innovative Research Inc Supplier No. IHUB2M1MG
Human Beta-2 Microglobulin (B2M) Purified 1mg - Human ATP5PB Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species Human Format Lyophilized Gene Symbol ATP5PB Gene Alias ATP5PB, PIG47, PIG47, ATP5F1 Protein Accession Number NP_001679 Nucelotide Accession Number NM_001688 Molecular Weight 28.9 kDa Storage Conditions -20C.