Learn More
Innovative Research Inc Hmn C-ReAct Prtn Kit
Supplier:  Innovative Research Inc IHUCRPKT
Encompass_Preferred
Human C-Reactive Protein ELISA Kit - The Human C-Reactive Protein ELISA Kit employs a quantitative enzyme immunoassay technique that measures C-Reactive Protein (CRP) in Human samples. Strip well format. Reagents for up to 96 tests. It is designed for detection of CRP in human plasma, serum, milk, urine, saliva, CSF, cell lysate, and cell culture samples. This assay employs a quantitative sandwich enzyme immunoassay technique that measures human CRP in less than 4 hours. A murine antibody specific for human CRP has been pre-coated onto a 96-well microplate with removable strips. CRP in standards and samples is sandwiched by the immobilized antibody and a biotinylated polyclonal antibody specific for human CRP, which is recognized by a streptavidin-peroxidase (SP) conjugate. All unbound material is washed away and a peroxidase enzyme substrate is added. The color development is stopped and the intensity of the color is measured.
Catalog No. 50-274-4341