Learn More
Innovative Research Inc Hmn CDS1 Prtn Lysate 20ug
Catalog No. 502744347
Encompass_Preferred
Human CDS1 Protein Lysate 20ug - Human CDS1 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species Human Format Lyophilized Gene Symbol CDS1 Gene Alias CDS1, CDS, CDS Protein Accession Number NP_001254 Nucelotide Accession Number NM_001263 Molecular Weight 53.1 kDa Storage Conditions -20C
Catalog No. 50-274-4347 Supplier Innovative Research Inc Supplier No. IHUCDS1PLLY20UG
Add to Cart
Add to Cart
Human CDS1 Protein Lysate 20ug - Human CDS1 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species Human Format Lyophilized Gene Symbol CDS1 Gene Alias CDS1, CDS, CDS Protein Accession Number NP_001254 Nucelotide Accession Number NM_001263 Molecular Weight 53.1 kDa Storage Conditions -20C