Learn More
Innovative Research Inc Hmn CLP1 Prtn Lysate 20ug
Catalog No. 502744344
Encompass_Preferred
Human CLP1 Protein Lysate 20ug - Human CLP1 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species HumanFormat Lyophilized Gene Symbol CLP1Gene Alias CLP1, hClp1, HEAB Protein Accession Number NP_006822Nucelotide Accession Number NM_006831Molecular Weight 47.6 kDa Storage Conditions -20C
Catalog No. 50-274-4344 Supplier Innovative Research Inc Supplier No. IHUCLP1PLLY20UG
Human CLP1 Protein Lysate 20ug - Human CLP1 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species HumanFormat Lyophilized Gene Symbol CLP1Gene Alias CLP1, hClp1, HEAB Protein Accession Number NP_006822Nucelotide Accession Number NM_006831Molecular Weight 47.6 kDa Storage Conditions -20C