Learn More
Proteintech Group Inc Hmn E-selectin /CD62E ELISA
SDP
Supplier:  Proteintech Group Inc KE0016996T
Encompass_Preferred
E-selectin, also known as ELAM-1 or CD62E, is a member of the selectin family of adhesion molecules that also include L-selectin and P-selectin. E-selectin is an inducible endothelial cell surface molecule. Its expression on endothelial cells is transcriptionally upregulated by various proinflammatory substances such as IL-1, TNFα and lipopolysaccharide (LPS). Besides, it can be secreted by endothelial cells, and can be detected in a soluble form (sE-selectin) in serum. During inflammation, E-selectin plays an important part in recruiting leukocytes to the site of injury.
Catalog No. 50-197-4123