Learn More
ACROBiosystems Human Fc gamma RI / CD64 Protein, His Tag (MALS verified) | 1mg
Supplier:  ACROBiosystems FCAH52H11MG
Encompass_Preferred
Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
Catalog No. NC1827207