Learn More
Abcam Human Factor IX/PTC ELISA Kit, 96 Tests
Supplier:  Abcam AB30030796TESTS
Encompass_Preferred
Human Factor IX/PTC ELISA Kit is a single-wash 90-min Simplestep used to quantify Human Factor IX/PTC with a sensitivity of 15.9 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10x96 plates for bulk orders and 384-well plate for higher throughput - Design your own immunoassay: we also offer the conjugation-ready antibody pair
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2197898