Learn More
Innovative Research Inc Hmn Fctr Beta XIIa Pur 100ug
Catalog No. 502744329
Encompass_Preferred
Human Factor Beta XIIa Purified 100ug - Human Factor Beta XIIa Purified from Innovative Research has been activated from Human Factor XII with Dextran Sulfate, then purified to remove the activator. This is a frozen liquid buffered in 4 mM sodium acetate-HCl/ 0.15 M NaCl/ pH 5.3,. Complete activation was observed by SDS-PAGE analysis. More Details Source Human Plasma Purification Prepared from Human Factor XII using Dextran Sulfate, then purified to remove the activator Purity Complete activation observed by SDS-PAGE analysis Storage Conditions -60 to -80C
Catalog No. 50-274-4329 Supplier Innovative Research Inc Supplier No. IHUFXIIAB100UG
Human Factor Beta XIIa Purified 100ug - Human Factor Beta XIIa Purified from Innovative Research has been activated from Human Factor XII with Dextran Sulfate, then purified to remove the activator. This is a frozen liquid buffered in 4 mM sodium acetate-HCl/ 0.15 M NaCl/ pH 5.3,. Complete activation was observed by SDS-PAGE analysis. More Details Source Human Plasma Purification Prepared from Human Factor XII using Dextran Sulfate, then purified to remove the activator Purity Complete activation observed by SDS-PAGE analysis Storage Conditions -60 to -80C