Learn More
Innovative Research Inc Hmn Fctr XIII Pur 250ug
Catalog No. 502744326
Encompass_Preferred
Human Factor XIII Purified 250ug - Human Factor XIII Purified from Innovative Research has been purified from human plasma. This is a frozen liquid buffered in 50 mM Tris/ 100 mM NaCl/ 1 mM EDTA/ 10 u/mL aprotinin/ 20% glycerol/ pH 7.5, and a purity of greater than 94% as determined by SDS-PAGE analysis. Also known as Fibrin Stablilizing Factor, Factor XIII is an enzyme that crosslinks Fibrin. Factor XIII is activated to XIIIa by Thrombin. Once activated, FXIIIa catalyzes covalent bonds between fibrin moledules, forming a stable and elastic insoluble clot. More Details Source Human Plasma Purity 94% by SDS-PAGE Storage Conditions -60 to -80C
Catalog No. 50-274-4326 Supplier Innovative Research Inc Supplier No. IHUFXIII250UG
Human Factor XIII Purified 250ug - Human Factor XIII Purified from Innovative Research has been purified from human plasma. This is a frozen liquid buffered in 50 mM Tris/ 100 mM NaCl/ 1 mM EDTA/ 10 u/mL aprotinin/ 20% glycerol/ pH 7.5, and a purity of greater than 94% as determined by SDS-PAGE analysis. Also known as Fibrin Stablilizing Factor, Factor XIII is an enzyme that crosslinks Fibrin. Factor XIII is activated to XIIIa by Thrombin. Once activated, FXIIIa catalyzes covalent bonds between fibrin moledules, forming a stable and elastic insoluble clot. More Details Source Human Plasma Purity 94% by SDS-PAGE Storage Conditions -60 to -80C