Learn More
Innovative Research Inc Hmn Fctr XIIIa Pur 250ug
Catalog No. 502744325
Encompass_Preferred
Human Factor XIIIa Purified 250ug - Human Factor XIIIa Purified from Innovative Research has been produced from Human Factor XIII that has been cleaved with Immobilized Human alpha-Thrombin, then purified using chromatography to remove the Thrombin. This is a frozen liquid buffered in 50 mM Tris/ 100 mM NaCl/ 1 mM EDTA/ 10 u/mL aprotinin / 50 uM DTT/ 50% glycerol/ pH 7.5,. Factor XIII is activated to XIIIa by Thrombin (with Calcium as a cofactor). Factor XIIIa acts as a stablilzer for fibrin by catalyzing covalent bonds between fibrin molecules, leading to the development of a stable, elastic fibrin clot structure. A clot that has not been stabalized by FXIIIa is soluble, whereas a Factor XIIIa-stabilized clot is insoluble. More Details Source Human Plasma Purification Human Factor XIII was cleaved with resin-immobilized Human alpha-Thrombin, then purified using chromatography to remove the Thrombin. Storage Conditions -20C
Catalog No. 50-274-4325 Supplier Innovative Research Inc Supplier No. IHUFXIIIA250UG
Human Factor XIIIa Purified 250ug - Human Factor XIIIa Purified from Innovative Research has been produced from Human Factor XIII that has been cleaved with Immobilized Human alpha-Thrombin, then purified using chromatography to remove the Thrombin. This is a frozen liquid buffered in 50 mM Tris/ 100 mM NaCl/ 1 mM EDTA/ 10 u/mL aprotinin / 50 uM DTT/ 50% glycerol/ pH 7.5,. Factor XIII is activated to XIIIa by Thrombin (with Calcium as a cofactor). Factor XIIIa acts as a stablilzer for fibrin by catalyzing covalent bonds between fibrin molecules, leading to the development of a stable, elastic fibrin clot structure. A clot that has not been stabalized by FXIIIa is soluble, whereas a Factor XIIIa-stabilized clot is insoluble. More Details Source Human Plasma Purification Human Factor XIII was cleaved with resin-immobilized Human alpha-Thrombin, then purified using chromatography to remove the Thrombin. Storage Conditions -20C