Learn More
Mabtech Inc HMN IFN-Y ELISPOTPRO KIT ALP
Supplier:  Mabtech Inc 34202APW2
Encompass
The ELISpotPRO kit is Mabtech's most innovative product featuring pre-coated plates and one-step detection. The kit enables enumeration of cells secreting human IFN-γ and contains ELISpot plates pre-coated with monoclonal antibody, ALP-conjugated antibody for one-step detection and BCIP/NBT-plus substrate. A polyclonal activator for human T cells (mAb CD3-2) is supplied as a positive control for cytokine secretion. Pre-coated plates and one-step detection enable a more rapid assay with minimized variability.Enzyme-conjugated detection mAb (7-B6-1-ALP) •Positive control (anti-CD3 mAb) •Substrate (BCIP/NBT-plus) •2 Pre-coated white plates (mAb 1-D1K)
Catalog No. NC0673003