Learn More
Innovative Research Inc Hmn IgA Depleted Srm 25ml
Catalog No. 502744318
Encompass_Preferred
Human IgA Depleted Serum 25ml - Human IgA Depleted Serum from Innovative Research has been purified from normal human serum by affinity chromatography. This is a Frozen liquid. Prepared from serum using immobilized Peptide M. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including tissue culture and more.
Catalog No. 50-274-4318 Supplier Innovative Research Inc Supplier No. IHUIGADS25ML
Human IgA Depleted Serum 25ml - Human IgA Depleted Serum from Innovative Research has been purified from normal human serum by affinity chromatography. This is a Frozen liquid. Prepared from serum using immobilized Peptide M. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including tissue culture and more.