Learn More
Innovative Research Inc Hmn IgA1 MyelomaPla AffPur 5mg
Catalog No. 502744319
Encompass_Preferred
Human IgA1 from Myeloma Plasma Affinity Purified 5mg - Human IgA1 from Myeloma Plasma Affinity Purified from Innovative Research has been purified from human myeloma plasma by affinity chromatography. This is a Frozen liquid buffered in 0.01M Sodium Phosphate; 0.15M NaCl; 0.05% Sodium Azide; pH7.4 with a purity of 95% as determined by SDS-PAGE. More Details Species Human Target IgA1 Purification Method Affinity purified Purity 95% as determined by SDS-PAGE Analysis Source Human myeloma plasma Storage Conditions -70 C Additional Information No reaction by IEP to IgA2 antiserum. This product can be used as an isotype control in applications like ELISA and Western Blot. It can also be used in a wide range of in vitro research applications. Concentration may vary from lot to lot. Please refer to the data sheet included with your order or contact us in advance if you seek a specific concentration.
Catalog No. 50-274-4319 Supplier Innovative Research Inc Supplier No. IHUIGA1MYAP5MG
Human IgA1 from Myeloma Plasma Affinity Purified 5mg - Human IgA1 from Myeloma Plasma Affinity Purified from Innovative Research has been purified from human myeloma plasma by affinity chromatography. This is a Frozen liquid buffered in 0.01M Sodium Phosphate; 0.15M NaCl; 0.05% Sodium Azide; pH7.4 with a purity of 95% as determined by SDS-PAGE. More Details Species Human Target IgA1 Purification Method Affinity purified Purity 95% as determined by SDS-PAGE Analysis Source Human myeloma plasma Storage Conditions -70 C Additional Information No reaction by IEP to IgA2 antiserum. This product can be used as an isotype control in applications like ELISA and Western Blot. It can also be used in a wide range of in vitro research applications. Concentration may vary from lot to lot. Please refer to the data sheet included with your order or contact us in advance if you seek a specific concentration.