Learn More
Innovative Research Inc Hmn KNG Sngl Chn Pur HMW 1mg
Catalog No. 502744295
Encompass_Preferred
Human Kininogen Single Chain Purified High Molecular Weight 1mg - Human Kininogen Single Chain Purified High Molecular Weight from Innovative Research has been purified from human plasma. This is a frozen liqud buffered in 4 mM sodium acetate-HCl/ 0.15 M NaCl/ pH 5.3, and a purity of greater than 95% as determined by SDS-PAGE analysis. High Molecular Weight Kininogen is plasma protein that helps to initiate coagulation. HMW Kininogen is one of the four proteins that begin the contact activation/intrinsic coagulation pathway (along with Factor XI, Factor XII, and Prekallikrein). HMW Kininogen functions as a cofactor in activating Kallikrein and Factor XII, and is necessary for Factor XIIa to activate Factor XI, but is not itself enzymatically active. More Details Source Human Plasma Purity > 95% by SDS-PAGE Storage Conditions -60 to -80C
Catalog No. 50-274-4295 Supplier Innovative Research Inc Supplier No. IHUKNGSCHMW1MG
Human Kininogen Single Chain Purified High Molecular Weight 1mg - Human Kininogen Single Chain Purified High Molecular Weight from Innovative Research has been purified from human plasma. This is a frozen liqud buffered in 4 mM sodium acetate-HCl/ 0.15 M NaCl/ pH 5.3, and a purity of greater than 95% as determined by SDS-PAGE analysis. High Molecular Weight Kininogen is plasma protein that helps to initiate coagulation. HMW Kininogen is one of the four proteins that begin the contact activation/intrinsic coagulation pathway (along with Factor XI, Factor XII, and Prekallikrein). HMW Kininogen functions as a cofactor in activating Kallikrein and Factor XII, and is necessary for Factor XIIa to activate Factor XI, but is not itself enzymatically active. More Details Source Human Plasma Purity > 95% by SDS-PAGE Storage Conditions -60 to -80C