Learn More
Innovative Research Inc Hmn KNG Two Chn Pur HMW 1mg
Supplier:  Innovative Research Inc IHUKNGTCHMW1MG
Encompass_Preferred
Human Kininogen Two Chain Purified High Molecular Weight 1mg - Human Kininogen Two Chain Purified High Molecular Weight from Innovative Research has been prepared by Kallikrein digestion of Kininogen, which is then purified to remove trace Kallikrein, resulting in the two-chain, Kinin-free form of Kininogen. This is a frozen liquid buffered in 4 mM sodium acetate-HCl/ 0.15 M NaCl/ pH 5.3, and a purity of greater than 95% as determined by SDS-PAGE analysis (complete reduction when incubated with 2-mercaptoethanol). More Details Source Human Plasma Purification Kallikrein digestion of Kininogen, purified to remove trace Kallikrein (resulting in two-chain, Kinin-free Kininogen) Purity 95% by SDS-PAGE (complete reduction when incubated with 2-mercaptoethanol) Storage Conditions -60 to -80C
Catalog No. 50-274-4294