Learn More
Proteintech Group Inc Hmn Mammaglobin A ELISA 96well
SDP
Supplier:  Proteintech Group Inc KE0007196T
Encompass_Preferred
Mammaglobin A, also known as SCGB2A2, is a member of the secretoglobin superfamily. Mammaglobin A is expressed in normal mammary gland epithelium and is over-expressed in primary and metastatic human breast cancers, making it a specific molecular marker and a potential therapeutic target for breast cancer. Secreted Mammaglobin protein has been detected in the sera of some breast cancer patients and not in sera of patients with other tumor types, indicating Mammaglobin as a serum marker for breast cancer
Catalog No. 50-197-4037