Learn More
Innovative Research Inc Hmn MYCT1 Prtn Lysate 20ug
Catalog No. 502744293
Encompass_Preferred
Human MYCT1 Protein Lysate 20ug - Human MYCT1 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles.Additional Details Species Human Format Lyophilized Gene Symbol MYCT1 Gene Alias MYCT1, MTLC, MTLC Protein Accession Number NP_079383 Nucelotide Accession Number NM_025107 Molecular Weight 26.4 kDa Storage Conditions -20C
Catalog No. 50-274-4293 Supplier Innovative Research Inc Supplier No. IHUMYCT1PLLY20UG
Human MYCT1 Protein Lysate 20ug - Human MYCT1 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles.Additional Details Species Human Format Lyophilized Gene Symbol MYCT1 Gene Alias MYCT1, MTLC, MTLC Protein Accession Number NP_079383 Nucelotide Accession Number NM_025107 Molecular Weight 26.4 kDa Storage Conditions -20C