Learn More
TRIBIOSCIENCE INC Human Neurofilament-Light Chain (NF-L) ELISA Kit, 1 x 96-well plate
SDP
Catalog No. NC2886073
Encompass_Preferred
The Human Neurofilament-Light Chain (NF-L) ELISA is designed to quantitatively detect human NF-L levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.156 to 10 ng/mL.
Catalog No. NC2886073 Supplier TRIBIOSCIENCE INC Supplier No. TBS32101
The Human Neurofilament-Light Chain (NF-L) ELISA is designed to quantitatively detect human NF-L levels in CSF, brain tissue, serum, plasma, and other biological samples. The kit uses proprietary methods to combine samples and detections in one step, rather than complex traditional methods. The detection range is from 0.156 to 10 ng/mL.