Learn More
ACROBiosystems Human NPR1 (NPRA) Extracellular Domain Protein, Fc Tag, 100 μg (MALS Verified)
Catalog No. NC3733224
Encompass_Preferred
Recombinant human natriuretic peptide receptor A (NPR1/NPRA) extracellular domain fused to a human IgG1 Fc tag. Produced in HEK293 cells and verified by size-exclusion chromatography with multi-angle light scattering (SEC-MALS) for molecular mass and oligomeric state. Supplied as an unconjugated 100 μg preparation suitable for biophysical characterization, ligand binding studies, and antibody development workflows.
  • Verified by SEC-MALS for molecular mass and oligomeric state.
  • Expressed in HEK293 human cells to preserve native glycosylation.
  • Carries a C-terminal human IgG1 Fc tag for dimerization and affinity capture.
  • Calculated molecular weight 75.3 kDa; migrates approximately 90-120 kDa by reducing SDS-PAGE.
  • Reported purity approximately 95% by SDS-PAGE.
  • Supplied unconjugated in 100 μg aliquots; store at -80 °C for long-term stability.
Catalog No. NC3733224 Supplier ACROBiosystems Supplier No. NP1H5259100UG
Recombinant human natriuretic peptide receptor A (NPR1/NPRA) extracellular domain fused to a human IgG1 Fc tag. Produced in HEK293 cells and verified by size-exclusion chromatography with multi-angle light scattering (SEC-MALS) for molecular mass and oligomeric state. Supplied as an unconjugated 100 μg preparation suitable for biophysical characterization, ligand binding studies, and antibody development workflows.
  • Verified by SEC-MALS for molecular mass and oligomeric state.
  • Expressed in HEK293 human cells to preserve native glycosylation.
  • Carries a C-terminal human IgG1 Fc tag for dimerization and affinity capture.
  • Calculated molecular weight 75.3 kDa; migrates approximately 90-120 kDa by reducing SDS-PAGE.
  • Reported purity approximately 95% by SDS-PAGE.
  • Supplied unconjugated in 100 μg aliquots; store at -80 °C for long-term stability.