Learn More
Innovative Research Inc Hmn NR3C2 Prtn Lysate 20ug
Catalog No. 502744292
Encompass_Preferred
Human NR3C2 Protein Lysate 20ug - Human NR3C2 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species HumanFormat Lyophilized Gene Symbol NR3C2Gene Alias NR3C2, MCR, MLR, MR, NR3C2VIT Protein Accession Number NP_000892 Nucelotide Accession Number NM_000901 Molecular Weight 106.9 kDa Storage Conditions -20C
Catalog No. 50-274-4292 Supplier Innovative Research Inc Supplier No. IHUNR3C2PLLY20UG
Human NR3C2 Protein Lysate 20ug - Human NR3C2 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species HumanFormat Lyophilized Gene Symbol NR3C2Gene Alias NR3C2, MCR, MLR, MR, NR3C2VIT Protein Accession Number NP_000892 Nucelotide Accession Number NM_000901 Molecular Weight 106.9 kDa Storage Conditions -20C