Learn More
Innovative Research Inc Hmn PAI1 and uPA Cmplx Kit
Catalog No. 502744277
Encompass_Preferred
Human PAI-1 and Urokinase (uPA) Complex ELISA Kit - This kit is for the quantitative determination of the covalent complex of uPA and its inhibitor PAI-1 in human plasma, serum, urine, tissue extracts, or cell culture media. For RUO. Strip well format reagents for up to 96 tests. Human uPA in samples will bind to the capture antibody coated on the microtiter plate. Free and complexed enzyme will react with the capture antibody on the plate. After washing, polyclonal anti human PAI-1 primary antibody binds to PAI-1/uPA complex captured on the plate. Excess antibody is washed away and bound polyclonal antibody is then reacted with the secondary antibody conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. A standard calibration curve of PAI-1/uPA complex is prepared along with the samples to be measured. Color development is proportional to the concentration of PAI-1/uPA complex. Free uPA and PAI-1 will not be detected by this assay.
Catalog No. 50-274-4277 Supplier Innovative Research Inc Supplier No. IHUPAI1UPAKTCX
Human PAI-1 and Urokinase (uPA) Complex ELISA Kit - This kit is for the quantitative determination of the covalent complex of uPA and its inhibitor PAI-1 in human plasma, serum, urine, tissue extracts, or cell culture media. For RUO. Strip well format reagents for up to 96 tests. Human uPA in samples will bind to the capture antibody coated on the microtiter plate. Free and complexed enzyme will react with the capture antibody on the plate. After washing, polyclonal anti human PAI-1 primary antibody binds to PAI-1/uPA complex captured on the plate. Excess antibody is washed away and bound polyclonal antibody is then reacted with the secondary antibody conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. A standard calibration curve of PAI-1/uPA complex is prepared along with the samples to be measured. Color development is proportional to the concentration of PAI-1/uPA complex. Free uPA and PAI-1 will not be detected by this assay.