Learn More
Innovative Research Inc Hmn PAI1 Rcmb Act Btn lbl
Catalog No. 502744290
Encompass_Preferred
Human PAI-1 Recombinant Active Biotin Labeled 500 ug - Human PAI-1 Recombinant Active Biotin Labeled from Innovative Research has been recombinantly produced in E. coli and purified by affinity chromatography. This is a Frozen liquid buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6 and a purity of >95% as determined by SDS-PAGE. More Details Species Human Target PAI-1 Purification Method Affinity purified Purity >95% as determined by SDS-PAGE Analysis Source E. coli Storage Conditions -70C Additional Information Active human PAI-1 biotin labeled by iodoacetamide substitution at the N-terminal cysteine. This product can be used as a control in applications like ELISA and Western Blot. It can be used in a wide range of in vitro applications, including blocking/inhibition, fibrinolysis, plasminogen activation, receptor binding, in hypertension studies, for protein-protein interactions, and more.
Catalog No. 50-274-4290 Supplier Innovative Research Inc Supplier No. IHUPAI1ARBL500UG
Human PAI-1 Recombinant Active Biotin Labeled 500 ug - Human PAI-1 Recombinant Active Biotin Labeled from Innovative Research has been recombinantly produced in E. coli and purified by affinity chromatography. This is a Frozen liquid buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6 and a purity of >95% as determined by SDS-PAGE. More Details Species Human Target PAI-1 Purification Method Affinity purified Purity >95% as determined by SDS-PAGE Analysis Source E. coli Storage Conditions -70C Additional Information Active human PAI-1 biotin labeled by iodoacetamide substitution at the N-terminal cysteine. This product can be used as a control in applications like ELISA and Western Blot. It can be used in a wide range of in vitro applications, including blocking/inhibition, fibrinolysis, plasminogen activation, receptor binding, in hypertension studies, for protein-protein interactions, and more.