Learn More
Innovative Research Inc Hmn PAI1 Rcmb Stbl Mut 500ug
Catalog No. 502744283
Encompass_Preferred
Human PAI-1 Recombinant Stable Mutant 500 ug - Human PAI-1 Recombinant Stable Mutant from Innovative Research has been produced in E. coli and purified by affinity chromatography. This is a frozen liquid buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6 and a purity of >95% as determined by SDS-PAGE. Species Human Target PAI-1 Purification Method Affinity purified Purity >95% as determined by SDS-PAGE Analysis Source E. coli Storage Conditions -70C; Protect from light Additional Information Human PAI-1 has four mutations (K154T, Q319L, M354I and N150H) that combine to create stability in the labile molecule, "locking" it to its active conformation. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including affinity purification, blocking, fibrinolysis, hypertension studies, inhibition, plasminogen activation, protein-protein interactions, receptor binding, and more
Catalog No. 50-274-4283 Supplier Innovative Research Inc Supplier No. IHUPAI1RSM500UG
Human PAI-1 Recombinant Stable Mutant 500 ug - Human PAI-1 Recombinant Stable Mutant from Innovative Research has been produced in E. coli and purified by affinity chromatography. This is a frozen liquid buffered in 0.05M Sodium Phosphate; 0.1M NaCl; 1mM EDTA; pH 6.6 and a purity of >95% as determined by SDS-PAGE. Species Human Target PAI-1 Purification Method Affinity purified Purity >95% as determined by SDS-PAGE Analysis Source E. coli Storage Conditions -70C; Protect from light Additional Information Human PAI-1 has four mutations (K154T, Q319L, M354I and N150H) that combine to create stability in the labile molecule, "locking" it to its active conformation. This product can be used as a control in applications like ELISA and Western Blot. It can be used for a wide range of in vitro applications, including affinity purification, blocking, fibrinolysis, hypertension studies, inhibition, plasminogen activation, protein-protein interactions, receptor binding, and more