Learn More
STEMCELL Technologies EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit, For processing: 1 x 10^9 cells
SDP
Catalog No. NC1538771
Encompass
The EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit is designed to deplete CD25+ cells from fresh peripheral blood mononuclear cells or lysed leukapheresis samples. CD25+ cells are targeted with Tetrameric Antibody Complexes recognizing CD25 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Labeled cells remain in the tube while unlabeled cells are poured off. The CD25 antigen is expressed on regulatory T cells and activated T and B cells.
Catalog No. NC1538771 Supplier STEMCELL Technologies Supplier No. 17861
The EasySep™ Human Pan-CD25 Positive Selection and Depletion Kit is designed to deplete CD25+ cells from fresh peripheral blood mononuclear cells or lysed leukapheresis samples. CD25+ cells are targeted with Tetrameric Antibody Complexes recognizing CD25 and dextran-coated magnetic particles. Labeled cells are separated using an EasySep™ magnet without the use of columns. Labeled cells remain in the tube while unlabeled cells are poured off. The CD25 antigen is expressed on regulatory T cells and activated T and B cells.