Learn More
Innovative Research Inc Hmn Plasmin Immobilized 1ml
Catalog No. 502744268
Encompass_Preferred
Human Plasmin Immobilized 1ml - Human Plasmin Immobilized has been purified from Human Plasma using affinity chromatography then immobilized to an agarose support. This is a resin buffered in 0.1M HEPES; 0.1M NaCl; 0.02% Sodium Azide; pH 7.4 coupled and a purity of >95%. This product is useful activation of single-chain tPA and uPA to the two-chain form and conversion of plasminogen from its native Glu form to the Lys form by removing the first 77 amino acids. More Details Source Human Plasma Purification Chromatographically purified Purity >95% Storage Conditions 4 C
Catalog No. 50-274-4268 Supplier Innovative Research Inc Supplier No. IHUPLMIMM1ML
Human Plasmin Immobilized 1ml - Human Plasmin Immobilized has been purified from Human Plasma using affinity chromatography then immobilized to an agarose support. This is a resin buffered in 0.1M HEPES; 0.1M NaCl; 0.02% Sodium Azide; pH 7.4 coupled and a purity of >95%. This product is useful activation of single-chain tPA and uPA to the two-chain form and conversion of plasminogen from its native Glu form to the Lys form by removing the first 77 amino acids. More Details Source Human Plasma Purification Chromatographically purified Purity >95% Storage Conditions 4 C