Learn More
Innovative Research Inc Hmn SMOC2 Prtn Lysate 20ug
Catalog No. 502744255
Encompass_Preferred
Human SMOC2 Protein Lysate 20ug - Human SMOC2 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional DetailsSpecies Human Format Lyophilized Gene Symbol SMOC2 Gene Alias SMOC2, bA270C4A.1, bA37D8.1, dJ421D16.1, DTDP1, MST117, MSTP117, MSTP140, SMAP2 Protein Accession Number NP_071421 Nucelotide Accession Number NM_022138 Molecular Weight 48.6 kDa Storage Conditions -20C
Catalog No. 50-274-4255 Supplier Innovative Research Inc Supplier No. IHUSMOC2PLLY20UG
Human SMOC2 Protein Lysate 20ug - Human SMOC2 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional DetailsSpecies Human Format Lyophilized Gene Symbol SMOC2 Gene Alias SMOC2, bA270C4A.1, bA37D8.1, dJ421D16.1, DTDP1, MST117, MSTP117, MSTP140, SMAP2 Protein Accession Number NP_071421 Nucelotide Accession Number NM_022138 Molecular Weight 48.6 kDa Storage Conditions -20C