Learn More
Lonza Walkersville Human Stem Cell Nucleofector Starter Kit
Catalog No. NC0143997
Encompass
The Nucleofector 2b Device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. Due to significant diversity between different human stem cell clones, we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit. The Human Stem Cell Nucleofector Starter Kit is used to determine which one of our two Nucleofector Kits is best suited for transfecting a specific primary human stem cell clone in Nucleofector 2b Device. It is suitable for 18 optimization reactions.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette format
Catalog No. NC0143997 Supplier Lonza Walkersville Supplier No. VPH5002
The Nucleofector 2b Device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. Due to significant diversity between different human stem cell clones, we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit. The Human Stem Cell Nucleofector Starter Kit is used to determine which one of our two Nucleofector Kits is best suited for transfecting a specific primary human stem cell clone in Nucleofector 2b Device. It is suitable for 18 optimization reactions.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette format