Learn More
Innovative Research Inc Hmn TAS2R46 Prtn Lysate 20ug
Catalog No. 502744254
Encompass_Preferred
Human TAS2R46 Protein Lysate 20ug - Human TAS2R46 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species Human Format Lyophilized Gene Symbol TAS2R46 Gene Alias TAS2R46, T2R46, T2R54 Protein Accession Number NP_795368 Nucelotide Accession Number NM_176887 Molecular Weight 35.3 kDa Storage Conditions -20C
Catalog No. 50-274-4254 Supplier Innovative Research Inc Supplier No. IHUTAS2R46PLLY20UG
Human TAS2R46 Protein Lysate 20ug - Human TAS2R46 Protein Lysate 20ug from Innovative Research is provided as a Lyophilized powder. This is a Recombinant Protein Lysate produced in HEK293T cells. This protein lysate can be reconsituted using SDS Sample Buffer. Once reconsituted, aliquot and store any remaining material at -20C to avoid repeated freeze-thaw cycles. Additional Details Species Human Format Lyophilized Gene Symbol TAS2R46 Gene Alias TAS2R46, T2R46, T2R54 Protein Accession Number NP_795368 Nucelotide Accession Number NM_176887 Molecular Weight 35.3 kDa Storage Conditions -20C