Learn More
Innovative Research Inc Hmn Ttl Antithrombin III Kit
Catalog No. 502744357
Encompass_Preferred
Human Total Antithrombin III ELISA Kit - Human Total Antithrombin III ELISA Kit from Innovative Research is intended for the quantitative determination of total antithrombin III antigen in human plasma. Strip well format. Reagents for up to 96 tests.Human antithrombin III will bind to the capture antibody coated on the microtiter plate. Free and complexed antithrombin III will react with the antibody on the plate. After appropriate washing steps, polyclonal anti-human antithrombin III primary antibody binds to the captured protein. Excess primary antibody is washed away and bound antibody, which is proportional to the total antithrombin III present in the samples, reacts with the secondary antibody. Following an additional washing step, TMB substrate is used for color development at 450nm.
Catalog No. 50-274-4357 Supplier Innovative Research Inc Supplier No. IHUATIIIKTT
Human Total Antithrombin III ELISA Kit - Human Total Antithrombin III ELISA Kit from Innovative Research is intended for the quantitative determination of total antithrombin III antigen in human plasma. Strip well format. Reagents for up to 96 tests.Human antithrombin III will bind to the capture antibody coated on the microtiter plate. Free and complexed antithrombin III will react with the antibody on the plate. After appropriate washing steps, polyclonal anti-human antithrombin III primary antibody binds to the captured protein. Excess primary antibody is washed away and bound antibody, which is proportional to the total antithrombin III present in the samples, reacts with the secondary antibody. Following an additional washing step, TMB substrate is used for color development at 450nm.