Learn More
Innovative Research Inc Hmn Ttl Fctr II Prthrmbn Kit
Catalog No. 502744334
Encompass_Preferred
Human Total Factor II (Prothrombin) ELISA Kit - This kit is for the quantitative determination of total prothrombin antigen in human plasma. For research use only. Strip well format for up to 96 tests. Human prothrombin will bind to the capture antibody coated on the microtiter plate. Prothrombin, thrombin, and thrombin-antithrombin complex will react with the antibody on the plate. After appropriate washing steps, anti human prothrombin primary antibody binds to the captured protein. Excess primary antibody is washed away and bound antibody is reacted with the secondary antibody. Following an additional washing step, TMB substrate is used for color development at 450nm. A standard calibration curve is prepared along with the samples to be measured using dilutions of human prothrombin. Color development is proportional to the concentration of total prothrombin in the samples. More Details Detection Target Prothrombin Reactivity Human Range 0.25-100 ng/ml Sensitivity 0.078 ng/ml.
Catalog No. 50-274-4334 Supplier Innovative Research Inc Supplier No. IHUFIIKTT
Human Total Factor II (Prothrombin) ELISA Kit - This kit is for the quantitative determination of total prothrombin antigen in human plasma. For research use only. Strip well format for up to 96 tests. Human prothrombin will bind to the capture antibody coated on the microtiter plate. Prothrombin, thrombin, and thrombin-antithrombin complex will react with the antibody on the plate. After appropriate washing steps, anti human prothrombin primary antibody binds to the captured protein. Excess primary antibody is washed away and bound antibody is reacted with the secondary antibody. Following an additional washing step, TMB substrate is used for color development at 450nm. A standard calibration curve is prepared along with the samples to be measured using dilutions of human prothrombin. Color development is proportional to the concentration of total prothrombin in the samples. More Details Detection Target Prothrombin Reactivity Human Range 0.25-100 ng/ml Sensitivity 0.078 ng/ml.