Learn More
Innovative Research Inc Hmn Ttl Fctr XI Kit
Catalog No. 502744323
Encompass_Preferred
Human Total Factor XI ELISA Kit - This human coagulation Factor XI antigen assay is intended for the quantitative determination of total Factor XI antigen in human plasma and serum. For research use only. Strip well format. Reagents for up to 96 tests. Human Factor XI will bind to the capture antibody coated on the microtiter plate. Both Factor XI and XIa will react with the antibody on the plate. After appropriate washing steps, biotinylated primary antibody binds to the captured protein. Excess primary antibody is washed away and bound antibody is reacted with horseradish peroxidase conjugated streptavidin. TMB substrate is used for color development at 450nm. A standard calibration curve is prepared along with the samples to be measured using dilutions of human Factor XI. Color development is proportional to the concentration of Factor XI in the samples.
Catalog No. 50-274-4323 Supplier Innovative Research Inc Supplier No. IHUFXIKTT
Human Total Factor XI ELISA Kit - This human coagulation Factor XI antigen assay is intended for the quantitative determination of total Factor XI antigen in human plasma and serum. For research use only. Strip well format. Reagents for up to 96 tests. Human Factor XI will bind to the capture antibody coated on the microtiter plate. Both Factor XI and XIa will react with the antibody on the plate. After appropriate washing steps, biotinylated primary antibody binds to the captured protein. Excess primary antibody is washed away and bound antibody is reacted with horseradish peroxidase conjugated streptavidin. TMB substrate is used for color development at 450nm. A standard calibration curve is prepared along with the samples to be measured using dilutions of human Factor XI. Color development is proportional to the concentration of Factor XI in the samples.