Learn More
Innovative Research Inc Hmn uPA Rcmb LMW 1mg
Catalog No. 502744238
Encompass_Preferred
Human Urokinase (uPA) Recombinant Low Molecular Weight 1mg - Human Urokinase (uPA) Recombinant Low Molecular Weight from Innovative Research has been recombinantly produced in insect cells and purified using a combination of conventional and affinity chromatography. This is a frozen liquid buffered in 0.05M Sodium Acetate; 0.1M NaCl; 1mM EDTA; pH 5.0 and a purity of 95%. It is useful as a positive control for ELISA and western blot and and other applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. Purified from auto-catalytic digestion of recombinant human urokinase produced in insect cells. Active two-chain low molecular weight human urokinase contains the proteinase domain and is separated from the amino terminal fragment by cleavage at the Lys135-Lys136 bond.
Catalog No. 50-274-4238 Supplier Innovative Research Inc Supplier No. IHUUPALMWR1MG
Human Urokinase (uPA) Recombinant Low Molecular Weight 1mg - Human Urokinase (uPA) Recombinant Low Molecular Weight from Innovative Research has been recombinantly produced in insect cells and purified using a combination of conventional and affinity chromatography. This is a frozen liquid buffered in 0.05M Sodium Acetate; 0.1M NaCl; 1mM EDTA; pH 5.0 and a purity of 95%. It is useful as a positive control for ELISA and western blot and and other applications including cell differentiation, cell signaling, clotting assays, fibrinolysis, plasminogen activation, protein-protein interactions, receptor binding, thromboelastography, and thrombolysis. Purified from auto-catalytic digestion of recombinant human urokinase produced in insect cells. Active two-chain low molecular weight human urokinase contains the proteinase domain and is separated from the amino terminal fragment by cleavage at the Lys135-Lys136 bond.