Learn More
Innovative Research Inc Hmn VIP Kit
Catalog No. 502744236
Encompass_Preferred
Human VIP ELISA Kit - The Human VIP ELISA Kit is an ELISA for measuring VIP in serum, plasma and other biological fluids. Reagents for up to 96 tests.This ELISA Kit uses the Competitive-ELISA principle. The micro ELISA plate provided in this Kit has been pre-coated with Human VIP. During the reaction, Human VIP in samples or Standard competes with a fixed amount of Human VIP on the solid phase supporter for sites on the Biotinylated Detection Ab specific to Human VIP. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) are added to each microplate well and incubated. Then a TMB substrate solution is added to each well. The enzyme-substrate reaction is terminated by the addition of stop solution and the color change is measured spectrophotometrically at a wavelength of 450 2 nm. The concentration of Human VIP in the samples is then determined by comparing the OD of the samples to the standard curve.M
Catalog No. 50-274-4236 Supplier Innovative Research Inc Supplier No. IHUVIPKT
Human VIP ELISA Kit - The Human VIP ELISA Kit is an ELISA for measuring VIP in serum, plasma and other biological fluids. Reagents for up to 96 tests.This ELISA Kit uses the Competitive-ELISA principle. The micro ELISA plate provided in this Kit has been pre-coated with Human VIP. During the reaction, Human VIP in samples or Standard competes with a fixed amount of Human VIP on the solid phase supporter for sites on the Biotinylated Detection Ab specific to Human VIP. Excess conjugate and unbound sample or standard are washed from the plate, and Avidin conjugated to Horseradish Peroxidase (HRP) are added to each microplate well and incubated. Then a TMB substrate solution is added to each well. The enzyme-substrate reaction is terminated by the addition of stop solution and the color change is measured spectrophotometrically at a wavelength of 450 2 nm. The concentration of Human VIP in the samples is then determined by comparing the OD of the samples to the standard curve.M