Learn More
Innovative Research Inc Hmn Von Willebrand Fctr Kit
Catalog No. 502744231
Encompass_Preferred
Human Von Willebrand Factor ELISA Kit - This kit is for the quantitative determination of Human Von Willebrand Factor in biofluid samples, such as plasma and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . vWF present in samples reacts with an anti-Human vWF capture antibody coated and dried on a microtiter plate. After washing, an anti-vWF detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is proportional to the concentration of active vWF. A standard calibration curve is prepared along with the samples to be measured using dilutions of Human vWF. Human Range 1.56-100ng/mL Sensitivity 0.53ng/mL Storage Conditions Store all components at 4C For research use only.
Catalog No. 50-274-4231 Supplier Innovative Research Inc Supplier No. IHUVWFKT
Human Von Willebrand Factor ELISA Kit - This kit is for the quantitative determination of Human Von Willebrand Factor in biofluid samples, such as plasma and other biological fluids. This is a sandwich ELISA in strip well format with reagents for up to 96 tests . vWF present in samples reacts with an anti-Human vWF capture antibody coated and dried on a microtiter plate. After washing, an anti-vWF detection antibody conjugated to biotin binds to the captured analyte. Excess antibody is washed away and bound antibody is then reacted with avidin conjugated to horseradish peroxidase. TMB substrate is used for color development at 450nm. Color development is proportional to the concentration of active vWF. A standard calibration curve is prepared along with the samples to be measured using dilutions of Human vWF. Human Range 1.56-100ng/mL Sensitivity 0.53ng/mL Storage Conditions Store all components at 4C For research use only.