Learn More
Promocell GmbH PromoCell hMNC-PB-c 25 million Human Mononuclear Cells, derived from Peripheral Blood, single donor, cryopreserved
Supplier:  Promocell GmbH C12907
Encompass
PromoCell Mononuclear Cells (MNC) represent the lymphocyte and monocyte fraction isolated from umbilical cord blood or peripheral blood. They are prepared by optimized low-density gradient centrifugation, which effectively reduces the granulocyte content. Red blood cells are gently but thoroughly depleted by proprietary techniques instead of osmotic lysis. This results in ultra-pure mononuclear cells with superior viability and unchanged biological function. In addition, the cells do not clump after thawing, allowing for easy handling and reproducible results.Our hMNC are now also available from HLA-typed donors.Please note: Cryopreserved human mononuclear cells are not suitable for the in vitro generation of macrophages using our Macrophage Generation Media XF. Always use freshly isolated human mononuclear cells or human monocytes for this application.
Catalog No. 50-306-071