Learn More
Aviva Systems Biology Corp HMOX1 Antibody - N-terminal region (ARP45222_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45222_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HMOX1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Human HMOX1. Alternate names: HO-1, HSP32, bK286B10ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-58987