Learn More
Biomatik Corporation HMOX1 Polyclonal Antibody
Supplier:  Biomatik Corporation CAF50158
Encompass_Preferred
HMOX1 Polyclonal Antibody is a Human (Homo sapiens) Polyclonal Antibody to Heme Oxygenase (Decycling) 1. The HMOX1 Polyclonal Antibody has been validated for the following applications: ELISA, WB, IHC, IP, IF. Host: Rabbit. Species: Human (Homo sapiens). Immunogen: 1~267aa. Applications: ELISA, WB, IHC, IP, IF. Target Synonyms: Heme Oxygenase (Decycling) 1. Accession Number: P09601. Gene ID: 3162. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6503