Learn More
Aviva Systems Biology Corp HMOX2 Antibody - N-terminal region (AVARP13071_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp AVARP13071_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against HMOX2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HMOX2. Alternate names: HO-2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62631