Learn More
Apexbio Technology LLC HMP Linker 68858-21-9 100g
SDP
Catalog No. 5000097251
Encompass_Preferred
HMP Linker is an acid-sensitive linker used in solid-phase peptide synthesis (SPPS) It is designed to facilitate the covalent attachment of peptide carboxy-terminal residues to solid-phase resin supports and enables controlled cleavage of synthesized peptides under mildly acidic conditions thereby preserving the structural integrity of sensitive functional groups and peptide termini HMP Linker acts primarily by hydrolyzing upon acid catalysis which allows the intact release of peptides from resin supports Based on these properties HMP Linker holds research potential in peptide therapeutics synthesis and studies requiring mild peptide cleavage conditions
Catalog No. 50-000-97251 Supplier Apexbio Technology LLC Supplier No. A7041100000
HMP Linker is an acid-sensitive linker used in solid-phase peptide synthesis (SPPS) It is designed to facilitate the covalent attachment of peptide carboxy-terminal residues to solid-phase resin supports and enables controlled cleavage of synthesized peptides under mildly acidic conditions thereby preserving the structural integrity of sensitive functional groups and peptide termini HMP Linker acts primarily by hydrolyzing upon acid catalysis which allows the intact release of peptides from resin supports Based on these properties HMP Linker holds research potential in peptide therapeutics synthesis and studies requiring mild peptide cleavage conditions