Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences HMPA (2-Hydroxypropyl Methacrylate) 450 ML
SDP

Catalog No. 5024320
Encompass_Preferred

(Propylene glycol monomethacrylate)
C₇H₁₂O₃ F.W. 144.17
Solubility: 13%
Boiling point: 70°C/1mmHg

Methacrylate is a water miscible monomer. Inhibited with approx. 300ppm MEHQ.

Catalog No. 50-243-20 Supplier Electron Microscopy Sciences Supplier No. 16900
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(Propylene glycol monomethacrylate)
C₇H₁₂O₃ F.W. 144.17
Solubility: 13%
Boiling point: 70°C/1mmHg

Methacrylate is a water miscible monomer. Inhibited with approx. 300ppm MEHQ.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.