Learn More
Electron Microscopy Sciences HMPA Embedding Kit
SDP
Catalog No. 50980395
Encompass_Preferred
HPMA is a water-soluble resin used for cytochemical applications. Infiltration follows fixation of the tissue so there is no extraction of materials caused by dehydration in alcohols. Leduc, E.H. et al., (1965) J. Cell Biol., 26:137. Kit consists of: 450 ml HPMA, 15g Benzoyl Peroxide Paste
Catalog No. 50-980-395 Supplier Electron Microscopy Sciences Supplier No. 14220
HPMA is a water-soluble resin used for cytochemical applications. Infiltration follows fixation of the tissue so there is no extraction of materials caused by dehydration in alcohols. Leduc, E.H. et al., (1965) J. Cell Biol., 26:137. Kit consists of: 450 ml HPMA, 15g Benzoyl Peroxide Paste