Learn More
Medchemexpress LLC HMR 1556 | 223749-46-0 | MFCD12756323 | 99.95% | 411.44 g/mol | C17H24F3NO5S | 1 ML
SDP
Catalog No. 5000243472
Encompass_Preferred
HMR 1556 is a chromanol-derived IKs (slow delayed rectifier potassium current) blocker used in cardiac electrophysiology research. It displays low-nanomolar potency in isolated ventricular myocytes and is supplied in solution or solid forms for laboratory use.
  • Potent IKs blocker with low-nanomolar IC50s.
  • Chromanol derivative selective for the slowly activating potassium current.
  • Supplied as a 10 mM solution in DMSO or as solid material for flexible dosing.
  • High reported purity suitable for research applications.
  • Analytical documentation available: SDS, COA, NMR, HPLC, LCMS.
  • For research use only; not for human or clinical use.
Catalog No. 50-002-43472 Supplier Medchemexpress LLC Supplier No. HY10636910MM 1ML
HMR 1556 is a chromanol-derived IKs (slow delayed rectifier potassium current) blocker used in cardiac electrophysiology research. It displays low-nanomolar potency in isolated ventricular myocytes and is supplied in solution or solid forms for laboratory use.
  • Potent IKs blocker with low-nanomolar IC50s.
  • Chromanol derivative selective for the slowly activating potassium current.
  • Supplied as a 10 mM solution in DMSO or as solid material for flexible dosing.
  • High reported purity suitable for research applications.
  • Analytical documentation available: SDS, COA, NMR, HPLC, LCMS.
  • For research use only; not for human or clinical use.