Learn More
BUCHI HMS Support Rod
SDP
Catalog No. 05376252 Shop All BUCHI Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
05-376-252 Stainless Steel
1 options
Catalog No. 05-376-252 Supplier BUCHI Supplier No. 11085647
Add to Cart
Add to Cart
Support for clamps and overhead temperature probes. Helps keep round-bottom flasks upright during stirring/heating.
  • For use with HMS Hot Plate Stirrers
  • Clamp and probe support sold separately
Specifications
Type Support Rod
Material Stainless Steel